Orders LinkContactUsLinkComponent
Stopcocks, high vacuum
Stopcocks, high vacuum
Supplier:   SciLabware
Stopcocks, high vacuum
Supplier:  SciLabware
Certificates
Stopcocks, high vacuum
Stopcocks, high vacuum
3 Options Available Below
Product Details & Documents
Borosilicate glass stopcocks specially designed for high vacuum applications.
  • All side-arms are 100 mm in length
  • Keys and barrels are precision ground together in matched pairs for optimum performance
  • Each key and barrel are numbered to ensure they are always paired together
Product Family Options
with straight bore
Product Information
  • Hole Ø
    For tube O-Ø
    Availability
    Price
  • 3 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €
  • 4 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
with double bore
Product Information
  • Hole Ø
    For tube O-Ø
    Availability
    Price
  • 4 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €