ContactUsLinkComponent
Molecular model: DNA, Molymod®
Molecular model: DNA, Molymod®
  SPIRMKO-ACGT-53
undefined
Molecular model: DNA, Molymod®
  SPIRMKO-ACGT-53
  • Description:
    Molecular model of DNA with four bases
  • Pk:
    1
This molecular model can show how the bases (adenin, cytosin, guanin, thymin) are combined through triple and double hydrogen bonds.