Orders LinkContactUsLinkComponent
Stopcocks, straight bore
Stopcocks, straight bore
Supplier:   SciLabware
Stopcocks, straight bore
Supplier:  SciLabware
Certificates
Stopcocks, straight bore
3 Options Available Below
Product Details & Documents
Straight bore stopcock, made of borosilicat glass 3.3, with plain arm and interchangeable glass key.
  • Glass stopcocks which meet the specifications of DIN 12541
  • Leakage rate meets the requirements of DIN 12540
Delivery information: Supplied complete with key retaining device.
Product Family Options
Product Information
  • Hole Ø
    For tube O-Ø
    Availability
    Price
  • 1,5 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,60 €
    17,60 €
    1 * 1 ST
       17,60 €
  • 2,5 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,20 €
    18,20 €
    1 * 1 ST
       18,20 €
  • 4,5 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,90 €
    19,90 €
    1 * 1 ST
       19,90 €
Accessories
Product Information
  • Catalog #
    Hole Ø
    For
    Availability
    Price
  • Replacement keys, for 12,5 mm Ø
    Replacement keys, for 12,5 mm Ø
    BIBBSSD102K
    1,5 mm
    SSD series stopcocks
    Log in for availability
    1 * 1 ST - 27,40 €
Product Information
  • Catalog #
    Hole Ø
    For
    Availability
    Price
  • Replacement keys, for 14,5 mm Ø
    Replacement keys, for 14,5 mm Ø
    BIBBSSD104K
    4,0 mm
    SSD series stopcocks
    Log in for availability
    1 * 1 ST - 26,20 €