Orders LinkContactUsLinkComponent
Screwthread joints
Screwthread joints
Supplier:   SciLabware
Screwthread joints
Supplier:  SciLabware
Certificates
Screwthread joints
Screwthread joints
4 Options Available Below
Product Details & Documents
Pyrex®, borosilicate glass.
  • Resistance to thermal shock and chemical corrosion
  • Designed to use with the range of plastic screwcaps, silicone rubber rings and PTFE washers
  • Unbadged
It is used by glassblowers in the construction of laboratory glassware.
Product Family Options
Product Information
  • Hole Ø
    Thread
    Tube ext.Ø
    Availability
    Price
  • 7,5 mm
    13 mm
    11,1 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 57,70 €
    57,70 €
    1 * 10 ST
       57,70 €
  • 10,5 mm
    18 mm
    15,4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 104,00 €
    104,00 €
    1 * 10 ST
       104,00 €
  • 15,5 mm
    24 mm
    22,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 136,00 €
    136,00 €
    1 * 10 ST
       136,00 €
  • 19,7 mm
    28 mm
    24,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 143,00 €
    143,00 €
    1 * 10 ST
       143,00 €