Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
Supplier:   SciLabware
Separating funnels, Gilson
Supplier:  SciLabware
Certificates
Separating funnels, Gilson
Separating funnels, Gilson
6 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, with NS-PE stopper.
Gilson style separating funnel with long delivery stem for good control of the liquid/liquid interface. With interchangeable glass or PTFE key for grease free performance. Supplied with hollow glass stopper.
Product Family Options
With PTFE key
Product Information
  • Capacity
    Cone size
    Availability
    Price
  • 60 ml
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,10 €
    87,10 €
    1 * 1 ST
       87,10 €
  • 125 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,70 €
    93,70 €
    1 * 1 ST
       93,70 €
  • 250 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • 500 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 146,00 €
    146,00 €
    1 * 1 ST
       146,00 €
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 173,00 €
    173,00 €
    1 * 1 ST
       173,00 €
  • 2000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,00 €
    248,00 €
    1 * 1 ST
       248,00 €