Order Entry
Austria
ContactUsLinkComponent
Ammonium 1-pyrrolidinecarbodithioate, Millipore®
Ammonium 1-pyrrolidinecarbodithioate, Millipore®
Catalog # 548000-100
CAS Number:  
Ammonium 1-pyrrolidinecarbodithioate, Millipore®
Catalog # 548000-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :146.3 · 18.0 g/mol
EINECS:225-834-4
CAS Number:5108-96-3
Formula:C₅H₁₂N₂S₂
MDL Number:MFCD00012720