Ammonium 1-pyrrolidinecarbodithioate 98%
:5108-96-3
:
:C₅H₁₂N₂S₂
:MFCD00012720
:146.3 · 18.0 g/mol
:150…155 °C
:225-834-4
:GHS07
:Refrigerator
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:97.5 to 102.5 %
Frequently Bought Together