Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standard, 2-(4-Chlorophenoxy)isobutyric acid 10 µg/ml in methanol
  PROONE9240
 :  
Organic reference standard, 2-(4-Chlorophenoxy)isobutyric acid 10 µg/ml in methanol
  PROONE9240
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    882-09-7
  • Description:
    2-(4-Chlorophenoxy)isobutyric acid 10 µg/ml in methanol
  • Formula:
    C₁₀H₁₁ClO₃
  • Description:
    2-(4-Chlorophenoxy)isobutyric acid