


Palladium(II) nitrate (4 - 5% Pd) (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10102-05-3
:
Formula:N₂O₆Pd
MDL Number:MFCD00011169
:230.43 g/mol
Storage Temperature:Ambient
EINECS:233-265-8
UN:3264
ADR:8,II
:GHS03
:GHS05
:GHS07
Related Information
Frequently Bought Together