VWR® AVS TITRINORM, Zirconium Standard Solution, 1000 mg/L Zr in 5% Hydrochloric Acid with Hydrofluoric Acid (max. 1%), Standard for AAS
Supplier: VWR Chemicals
Certificates
About this item
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%
Frequently Bought Together