5-(Bromomethyl)-2,1,3-benzothiadiazole
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:65858-50-6
:
Formula:C₇H₅BrN₂S
MDL Number:MFCD00544836
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together