VWR® AVS TITRINORM, Europium Standard Solution, 1000 mg/L Eu in dilute Nitric Acid AVS TITRINORM Standard for AAS
Artikelnr. 86676.260
Lieferant: VWR Chemicals
CAS-Nummer:
Spezifikationen
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Environmentally Preferable:
- Element:Europium
- Symbol:Eu
Spezifikationen
Über diesen Artikel
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%