Calcium sulphate, Drierite™ 20-40 mesh for drying, without indicator
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum
CAS Number:7778-18-9
:
Formula:CaO₄S
MDL Number:MFCD00010912
Molecular Weight:136.14 g/mol
Melting Pt:1450 °C
Merck Index:13,01711
Storage Temperature:Ambient
EINECS:231-900-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white Irregular granules
- Water:≤0.3 %
- Particle size:20 to 40 mesh
- Composition:≥99.5 % (CaSO4) Typical
Specification Test Results
Frequently Bought Together