Calcium sulphate, Drierite™ 8 mesh for drying, without indicator
: Calcium sulphate (anhydrous) anhydrite, Gypsum
:7778-18-9
:
:CaO₄S
:MFCD00010912
:136.14 g/mol
:1450 °C
:13,01711
:Ambient
:231-900-3
- Environmentally Preferable:
- Appearance:White to off-white Irregular granules
- Water:≤0.3 %
- Particle size:8 mesh
- Composition:≥99.5 % (CaSO4) Typical
Frequently Bought Together