Standard conical ground joints sockets, Quickfit®
PYREX® borosilicate glass.
- Accurate taper and fine surface finish reduce possibility of joint 'seizure' or leakage
- Manufactured to an accuracy which surpasses the requirements of ISO 383
- Avoids the problem of the badge charring at high temperatures which can give an untidy appearance
Unbadged ground glass sockets free from any ceramic markings.
Total length: 100 mm
Frequently Bought Together