Standard conical ground joints cones, Quickfit®
Supplier: SciLabware
Certificates
About this item
PYREX® borosilicate glass.
- Resistance to chemical corrosion and thermal shock
- Manufactured to an accuracy which surpasses the requirements of ISO 383
- Avoids the problem of the badge charring at high temperatures which can give an untidy appearance
Unbadged ground glass cones free from any ceramic markings.
Total length: 100 mm
Frequently Bought Together