Automatic burettes, Pellet pattern, class AS
Burettes made of chemically resistant PYREX® borosilicate glass.
- Automatic zero facility
- Simple operation
- White enamel graduations
- 1,5 mm bore PTFE key stopcock
: Complies with ISO 385 and DIN 12700.
Product Family Options
- CapacityDivision