Order Entry
Austria
Orders LinkContactUsLinkComponent
Membrane filter, silver

Membrane filter, silver

Silver, 0,45 µm, 25 mm
Catalog # MILFAG4502550
Supplier:   Merck
Membrane filter, silver
Catalog # MILFAG4502550
Supplier:  Merck
Membrane filter, silver
Membrane filter, silver

Silver, 0,45 µm, 25 mm

Price
835,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

These silver membrane filter are suitable for a variety of applications and are commonly used for air monitoring and analysis of particulate contaminants. They are specified for collection and detection of particles such as silica, carbon black, coal tar products and coke oven emissions.

  • Pure silver filters for particulate monitoring
  • Highly resistant to thermal stress and aggressive chemicals
  • Low background for sensitive X-ray diffraction analysis

Used for air monitoring: asbestos, lead sulphide, crystalline and amorphous silica.

Specifications
  • Ø:
    25 mm
  • Pore size:
    0,45 µm
  • Type:
    Silver