ContactUsLinkComponent
Magnetic stirring bars, PTFE-coated, octagonal, with centre ring
Magnetic stirring bars, PTFE-coated, octagonal, with centre ring
Catalog # HECH42077008
undefined
Magnetic stirring bars, PTFE-coated, octagonal, with centre ring
Catalog # HECH42077008
Specifications
  • Ø:
    8 mm
  • Type:
    with centre ring
  • Length:
    51 mm
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
PTFE coated, magnetic stirring bars.
  • Pivot ring reduces surface contact and ensures stirrer is in optimum position in the vessel.