Orders LinkContactUsLinkComponent
Screw caps
Screw caps
Supplier:   VITLAB
Screw caps
Supplier:  VITLAB
Certificates
Screw caps
3 Options Available Below
Product Details & Documents
PFA, transparent
  • Especially suitable for use in trace analysis
  • Ideal for sensitive and valuable samples
  • High thermal stability, from –200 °C to +260 °C, autoclavable
  • Chemical inertness against nearly all chemicals
  • Easy to clean
For sealing all PFA containers with GL threads or buttress threads.
Product Family Options
Product Information
  • Thread
    Availability
    Price
  • S 28
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,90 €
    28,90 €
    1 * 1 ST
       28,90 €
  • GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,90 €
    5,90 €
    1 * 1 ST
       5,90 €
  • GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,60 €
    11,60 €
    1 * 1 ST
       11,60 €