Membrane filters, PORAFIL® RC
Supplier: MACHEREY-NAGEL
Certificates
About this item
Regenerated cellulose membranes are used for filtration of solvent mixtures and ultrapurification and degassing of HPLC eluents.
- Resistant to most organic solvents
Frequently Bought Together