Sample Loops for Rheodyne® Valves
Supplier: Agilent
Certificates
About this item
These PEEK sample loops are inert to most organic solvents. The stainless-steel sample loops are not interchangeable due to the change in port angle.
- Stainless steel loops are square cut and free of burrs for a flush connection
- Flexible PEEK loops have a clean, straight cut for low dead volume connections
Accuracy of large PEEK loops is ±14%, intermediate loops ±21%, small loops ±65%. Accuracy of large metal loops is ±5%, intermediate loops ±10%, small loops ±30%
Frequently Bought Together