About this item
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
- Can be sterilised by EO, gamma, or autoclave.
Type APFA: Retains fine particles with good efficiency, even at high flow rates, used for monitoring wastewater and collecting suspended particles in gases, also have biochemical applications like collection of cells and filtration of protein or nucleic acid precipitates.
Type APFB: Higher mechanical strength when wet and greater loading capacity than type APFA filters, used for liquid clarification, quantification of solids in suspensions of fine particles, and scintillation counting.
Type APFC: Greater retention, especially for fine particulate and microorganism removal than type APFA filters , used for determining total suspended solids in drinking water and filtering proteins or nucleic acid TCA precipitates and for collecting cells and microorganisms.
Type APFD and APFF: APFD are thick filter with high flow and low retention, used for clarifying suspensions containing particulates >1,0 µm. APFF used for filtering extremely fine precipitates such as protein, nucleic acids, or serum precipitates, recommended for USEPA method 1311 for TCLP analysis.
Type AP40: Recommended for standard methods total suspended solids 2540D, recommended for USEPA method 1311 for TCLP analysis. Maintains structural integrity without weight loss when ignited to 550 °C after sample filtration, used for determining volatile suspended matter in wastewater and industrial effluents.
Specifications
- Catalog No:
- MILFAPFD09050
- MILFAPFF04700
- MILFAPFC02500
- MILFAP4003705
- MILFAPFF09050
- MILFAP4014250
- MILFAP4009000
- MILFAP4007000
- MILFAPFB02500
- MILFAP408X105
- MILFAPFC03700
- MILFAPFF14250
- MILFAPFF02500
- MILFAPFF12450
- MILFAP4004700
- MILFAPFB15050
- MILFAP4002405
- 512-0243
- MILFAPFA04700
- MILFAPFA09050
- MILFAPFB04700
- MILFAPFB03700
- 512-0244
- MILFAP4001000
- MILFAPFC09050
- MILFAPFD02500
- MILFAP4002500
- MILFAPFD04700
- Weight:
- 120 g/m²
- 75 g/m²
- 52 g/m²
- 65 g/m²
- 75 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 140 g/m²
- 65 g/m²
- 52 g/m²
- 75 g/m²
- 75 g/m²
- 75 g/m²
- 65 g/m²
- 140 g/m²
- 65 g/m²
- 65 g/m²
- 55 g/m²
- 55 g/m²
- 140 g/m²
- 140 g/m²
- 52 g/m²
- 65 g/m²
- 52 g/m²
- 120 g/m²
- 65 g/m²
- 120 g/m²
- Thickness:
- 470 µm
- 380 µm
- 240 µm
- 475 µm
- 380 µm
- 475 µm
- 475 µm
- 475 µm
- 700 µm
- 475 µm
- 240 µm
- 380 µm
- 380 µm
- 380 µm
- 475 µm
- 700 µm
- 475 µm
- 475 µm
- 230 µm
- 230 µm
- 700 µm
- 700 µm
- 240 µm
- 475 µm
- 240 µm
- 470 µm
- 475 µm
- 470 µm
- Particle retention:
- 2,7 µm
- 0,7 µm
- 1,2 µm
- 0,7 µm
- 1,0 µm
- 1,2 µm
- 0,7 µm
- 0,7 µm
- 0,7 µm
- 1,0 µm
- 1,6 µm
- 1,6 µm
- 1,0 µm
- 1,0 µm
- 1,2 µm
- 1,2 µm
- 2,7 µm
- 2,7 µm
- Pore size:
- 2,7 µm
- 0,7 µm
- 1,2 µm
- 0,7 µm
- 0,7 µm
- 0,7 µm
- 0,7 µm
- 0,7 µm
- 1,0 µm
- 0,7 µm
- 0,7 µm
- 0,7 µm
- 0,7 µm
- 0,7 µm
- 1,6 µm
- 1,6 µm
- 1,0 µm
- 1,0 µm
- 1,2 µm
- 0,7 µm
- 1,2 µm
- 2,7 µm
- 0,7 µm
- 2,7 µm
- Water flow rate:
- 2,7 ml/min/cm²
- 1,4 ml/min/cm²
- 1,2 ml/min/cm²
- 6 ml/min/cm²
- 1,4 ml/min/cm²
- 6 ml/min/cm²
- 6 ml/min/cm²
- 6 ml/min/cm²
- 2,2 ml/min/cm²
- 6 ml/min/cm²
- 1,2 ml/min/cm²
- 1,4 ml/min/cm²
- 1,4 ml/min/cm²
- 1,4 ml/min/cm²
- 6 ml/min/cm²
- 2,2 ml/min/cm²
- 6 ml/min/cm²
- 6 ml/min/cm²
- 5 ml/min/cm²
- 5 ml/min/cm²
- 2,2 ml/min/cm²
- 2,2 ml/min/cm²
- 1,2 ml/min/cm²
- 6 ml/min/cm²
- 1,2 ml/min/cm²
- 2,7 ml/min/cm²
- 6 ml/min/cm²
- 2,7 ml/min/cm²
- Airflow rate:
- 139 L/min/cm² at 10 psi
- 18,6 L/min/cm² at 10 psi
- 41,2 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 18,6 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 23,4 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 41,2 L/min/cm² at 10 psi
- 18,6 L/min/cm² at 10 psi
- 18,6 L/min/cm² at 10 psi
- 18,6 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 23,4 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 67,5 L/min/cm² at 10 psi
- 67,5 L/min/cm² at 10 psi
- 23,4 L/min/cm² at 10 psi
- 23,4 L/min/cm² at 10 psi
- 41,2 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 41,2 L/min/cm² at 10 psi
- 139 L/min/cm² at 10 psi
- 44,5 L/min/cm² at 10 psi
- 139 L/min/cm² at 10 psi
- Max. temperature:
- 500 °C
- 500 °C
- 500 °C
- 550 °C
- 500 °C
- 550 °C
- 550 °C
- 550 °C
- 500 °C
- 550 °C
- 500 °C
- 500 °C
- 500 °C
- 500 °C
- 550 °C
- 500 °C
- 550 °C
- 550 °C
- 500 °C
- 500 °C
- 500 °C
- 500 °C
- 500 °C
- 550 °C
- 500 °C
- 500 °C
- 550 °C
- 500 °C