Order Entry
Austria
ContactUsLinkComponent
Cation exchange chromatography columns, TSK-GEL® SP-2SW
Cation exchange chromatography columns, TSK-GEL® SP-2SW
Catalog # TOYO0007165
Supplier:  TOSOH Bioscience
undefined
Cation exchange chromatography columns, TSK-GEL® SP-2SW
Catalog # TOYO0007165
Supplier:  TOSOH Bioscience

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    125 Å
  • Description:
    SP-2SW
  • Length:
    250 mm
  • Column hardware:
    Stainless steel
  • Pk:
    1

Specifications

About this item

TSK-GEL® SP-2SW are silica based stainless steel column packed with spherical silica beads which are surface modified with sulfopropyl strong cation exchange groups. Designed for the separation and analysis of small protein, peptides, nucleosides and other biologically active molecules.