Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Storage Boxes, 96 Well

   Sustainable Options Available
Supplier:   Greiner Bio-One
Storage Boxes, 96 Well
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates

2 Options Available Below

Product Details & Documents

The storage boxes are made of polycarbonate while the individual tubes are made of biologically inert polypropylene.

  • Comes with a coding card, which enables proper storage of samples
  • Temperature resistance of the polypropylene tubes from −80 °C to +121 °C provides for a broad range of applications
Specifications for Product Family

Specifications

  • Catalog No:
  • GREI975502BE
  • GREI975502_1
  • 391-3726
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (241002); ISO 50001 (241002)

Specifications

Product Family Options

Product Information

  • Capacity
    Material
    Availability
    Price
  • GREI975502BE
    Storage box with ID card and lid, without tubes, non sterile
    Storage box with ID card and lid, without tubes, non sterile
    1,2 ml in each tube
    PC
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    GREI975502BE
    Environmentally Preferable
    Environmental certification
  • GREI975502_1
    Storage box with ID card and lid, without tubes, non sterile
    Storage box with ID card and lid, without tubes, non sterile
    1,2 ml in each tube
    PC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,40 €
    51,40 €
    1 * 1 ST
       51,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    GREI975502_1
    Environmentally Preferable
    Environmental certification
  • 391-3726
    Storage box with lid and ID card, inserted with 96 well PP vessels, non sterile
    Storage box with lid and ID card, inserted with 96 well PP vessels, non sterile
    1,2 ml in each tube
    PC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,90 €
    77,90 €
    1 * 1 ST
       77,90 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-3726
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact

PCR Tubes for 96 well storage boxes

Product Information

  • Catalog #
    Colour
    Description
    Capacity
    Availability
    Price
  • Natural
    Tube strips,with attached 8-cap strips, non sterile, Ø 8,5 mm, height 44 mm
    1,3 ml
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 274,00 €
    274,00 €
    1 * 1.000 ST
       274,00 €

Product Information

  • Catalog #
    Colour
    Description
    Capacity
    Availability
    Price
  • Natural
    Single tubes, non sterile, without cap, Ø 8,5 mm, height 44 mm
    1,3 ml
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 74,80 €
    74,80 €
    1 * 1.000 ST
       74,80 €