ContactUsLinkComponent
Upright microscopes, BA210 Elite and BA310 Elite
Upright microscopes, BA210 Elite and BA310 Elite
Supplier:  MOTIC
Certificates
undefined
Upright microscopes, BA210 Elite and BA310 Elite
Supplier:  MOTIC
Specifications
  • Catalog No:
  • 630-1826
  • 630-1825
  • MOTI1100100402966
  • MOTI1100100403221
  • Model:
  • BA310 Elite
  • BA310 Elite
  • BA310 Elite
  • BA310 Elite
  • Observation method:
  • Brightfield
  • Brightfield
  • Brightfield
  • Brightfield
  • Head:
  • 30° inclined, 360° rotating
  • 30° inclined, 360° rotating
  • 30° inclined, 360° rotating
  • 30° inclined, 360° rotating
  • Eyepieces:
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • Nosepiece:
  • Quintuple, reversed
  • Quintuple, reversed
  • Quintuple, reversed
  • Quintuple, reversed
  • Stage:
  • 180×170 mm (movement: 80×55 mm)
  • 180×170 mm (movement: 80×55 mm)
  • 180×170 mm (movement: 80×55 mm)
  • 180×170 mm (movement: 80×55 mm)
  • Condenser:
  • Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
  • Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
  • Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
  • Abbe N.A. 0,90/1,25 with iris diaphragm and slider slot
  • Focusing system:
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Illumination:
  • Halogen interchangeable with LED module
  • Halogen interchangeable with LED module
  • Halogen interchangeable with LED module
  • Halogen interchangeable with LED module
Specifications
Objectives
Product Information
Frequently Bought Together