Erlenmeyer flasks, narrow or wide neck, beaded rim
: Kavalier
Made of borosilicate glass 3.3., clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.
Frequently Bought Together