About this item
These centrifugal filters are disposable ultrafiltration non sterile devices used for purifying, concentrating, and desalting biological samples in the 2 to 15 ml volume range.
- For use with samples containing high levels of solutes
- Unique inverse flow mode of operation with large dead stop
- Fast sample processing
- Fits standard swinging-bucket rotor for 50 ml tubes
They may also be used for filtration applications. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 ml centrifuge tubes. They are easy to use and offer a high flow rate. This devices consist of a sample container with a twistlock cap, a filtrate collector containing a low-adsorptive, Ultracel® regenerated cellulose membrane and an air-seal cap for sample isolation.
Application includes concentration and purification of particle-laden solutions, solutions with high solute concentration, separation of low MW solutes from fermentation broths, cell culture media and lysates.
Specifications
- Catalog No:
- MILF4307
- MILF4303
- MILF4311
- MILF4302
- Filtration area:
- 2,84 cm²
- 2,84 cm²
- 2,84 cm²
- 2,84 cm²
- Max. RCF:
- 1500 ×g
- 3000 ×g
- 3000 ×g
- 3000 ×g
- Dead volume:
- 500 µl
- 500 µl
- 500 µl
- 500 µl
- Ø×H:
- 28,1×135 mm
- 28,1×135 mm
- 28,1×135 mm
- 28,1×135 mm
- Sample volume:
- 15000 µl
- 15000 µl
- 15000 µl
- 15000 µl