Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PPL
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PPL
Supplier:  VARIAN
Certificates
9 Options Available Below
Product Details & Documents
Bond Elut™ PPL is a styrene-divinylvenzene (SDVB) polymer that has been modified with a proprietary non-polar surface. PPL will retain even the most polar classes of analytes, including phenols. The large particle size allows ease of flow for viscous or particulate rich water samples, while the high surface area and strong hydrophobicity ensure reproducible extractions with high recoveries upon elution. Bond Elut™ PPL is suitable for EPA Method 528 ‘Determination of Phenols in Drinking Water by SPE and Capillary GC/MS’.
  • Modified styrene-divinylbenzene polymer
  • Large particle size allows fast extraction speeds
  • High surface area and capacity for polar analytes
Specifications for Product Family
Specifications
  • Catalog No:
  • 551-0331
  • VARI12105003
  • 551-0332
  • 551-0333
  • VARI12105006
  • 551-0400
  • VARI12102148
  • 551-0401
  • VARI12256087
  • Surface area:
  • 600 m²/g
  • 600 m²/g
  • 600 m²/g
  • 600 m²/g
  • 600 m²/g
  • 600 m²/g
  • 600 m²/g
  • 600 m²/g
  • 600 m²/g
Specifications
Product Family Options
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 551-0331
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 270,07 €
    270,07 €
    1 * 100 ST
       270,07 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0331
    Surface area
    600 m²/g
  • VARI12105003
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 345,77 €
    345,77 €
    1 * 100 ST
       345,77 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12105003
    Surface area
    600 m²/g
  • 551-0332
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 229,15 €
    229,15 €
    1 * 50 ST
       229,15 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0332
    Surface area
    600 m²/g
  • 551-0333
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 317,13 €
    317,13 €
    1 * 50 ST
       317,13 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0333
    Surface area
    600 m²/g
  • VARI12105006
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 561,63 €
    561,63 €
    1 * 50 ST
       561,63 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12105006
    Surface area
    600 m²/g
  • 551-0400
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 336,57 €
    336,57 €
    1 * 30 ST
       336,57 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0400
    Surface area
    600 m²/g
  • VARI12102148
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    3 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1 053,69 €
    1 053,69 €
    1 * 50 ST
       1 053,69 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102148
    Surface area
    600 m²/g
  • 551-0401
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 513,55 €
    513,55 €
    1 * 30 ST
       513,55 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0401
    Surface area
    600 m²/g
  • VARI12256087
    Straight barrel cartridge
    Straight barrel cartridge
    PPL
    150 Å
    125 µm
    60 ml
    5 g
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 1 411,74 €
    1 411,74 €
    1 * 16 ST
       1 411,74 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256087
    Surface area
    600 m²/g