Solid phase extraction cartridges, Bond Elut™ PH
Anbieter: VARIAN
About this item
Bond Elut™ PH is a non-polar bonded silica material which exhibits a different selectivity to alkyl or aliphatic functionalised phases such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.
- Added selectivity compared to other non-polar sorbents
- Enhanced retention of planar, conjugated organic molecules
- Similar polarity to C8
11 Options Available Below
Product Details & Documents
Bond Elut™ PH is a non-polar bonded silica material which exhibits a different selectivity to alkyl or aliphatic functionalised phases such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.
- Added selectivity compared to other non-polar sorbents
- Enhanced retention of planar, conjugated organic molecules
- Similar polarity to C8
Specifications for Product Family
Specifications
- Catalog No:
- VARI12113005
- VARI14113005
- VARI12113031
- VARI12102062
- 551-0303
- 551-0318
- VARI12256004
- VARI14102062
- VARI14102005
- VARI14102032
- VARI14256004
- Carbon load:
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- 10,7%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
- DescriptionPhasePore sizeParticle SizeCapacityBed weightVerfügbarkeitPreis
- Specifications:
Mehr
Specifications
Cat. No.VARI12113005Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.VARI14113005Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.VARI12113031Carbon load10,7%EndcappedYesSurface area500 m²/g
Straight barrel cartridges
Product Information
- DescriptionPhasePore sizeParticle SizeCapacityBed weightVerfügbarkeitPreis
- Specifications:
Mehr
Specifications
Cat. No.VARI12102062Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.551-0303Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.551-0318Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.VARI12256004Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.VARI14102062Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.VARI14102005Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.VARI14102032Carbon load10,7%EndcappedYesSurface area500 m²/g - Specifications:
Mehr
Specifications
Cat. No.VARI14256004Carbon load10,7%EndcappedYesSurface area500 m²/g



