Order Entry
Austria
ContactUsLinkComponent
 
Solid phase extraction cartridges, Bond Elut™ PCB
Catalog # VARI12105032
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PCB
Catalog # VARI12105032
Supplier:  VARIAN

Specifications

  • Capacity:
    3 ml
  • Description:
    LRC cartridge
  • Surface area:
    500 m²/g
  • Bed weight:
    1 g
  • Phase:
    PCB
  • Pk:
    50

Specifications

About this item

Bond Elut™ PCB is a specially designed sorbent which allows for the facile extraction of polychlorinated biphenyl (PCB) compounds from a variety of matrices. Desired analytes can be loaded and eluted using a simple, single solvent method prior to analysis by GC/ECD.

  • Optimised bed mass affording excellent extraction reproducibility
  • Special dual-phase enhances PCB selectivity
  • All extractions can be completed with one solvent to simplify procedures