Solid phase extraction cartridges, Bond Elut™ PBA
Supplier: VARIAN
Certificates
About this item
PBA is a unique sorbent consisting of a phenylboronic acid covalently linked to a silica gel surface. Covalent processes result in a higher retention to the sorbent surface than other mechanisms, resulting in superior clean-up. The boronate group has a high specificity for cis-diol containing compounds such as catechols, nucleic acids, low molecular weight proteins and carbohydrates.
- High specificity for cis-diol compounds
- Very effective in separating DNA and RNA
Bond Elut™ PBA can also be used to retain oligomeric materials such as polyethylene glycols (PEGs) from a variety of aqueous or biological matrices.
Specifications
- Catalog No:
- VARI12102127
- VARI12102019
- VARI12102105
- 551-0344
- VARI12102018
- VARI12105033
- Carbon load:
- 7,9%
- 7,9%
- 7,9%
- 7,9%
- 7,9%
- 7,9%
- Endcapped:
- No
- No
- No
- No
- No
- No
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specifications
Frequently Bought Together