Solid phase extraction cartridges, EnvirElut™
About this item
EnvirElut™ sorbents are specially designed for the extraction of a wide range of compounds from aqueous matrices. EnvirElut™ PAH and Pesticides are available in standard SPE straight barrel cartridges, which can be used on conventional vacuum manifolds such as the Vac Elut SPS 24.
- Extreme purity offers cleanliness in extract
- High capacity allows for the processing of large sample volumes
- Broad compound specificity
6 Options Available Below
Product Details & Documents
EnvirElut™ sorbents are specially designed for the extraction of a wide range of compounds from aqueous matrices. EnvirElut™ PAH and Pesticides are available in standard SPE straight barrel cartridges, which can be used on conventional vacuum manifolds such as the Vac Elut SPS 24.
- Extreme purity offers cleanliness in extract
- High capacity allows for the processing of large sample volumes
- Broad compound specificity
Specifications for Product Family
Specifications
- Catalog No:
- VARI12102158
- VARI12272005
- VARI12272004
- VARI12272001
- VARI12272011
- VARI12272020
- Carbon load:
- 18,0%
- 18,0%
- 18,0%
- 18,0%
- 18,0%
- Endcapped:
- No
- No
- No
- No
- No
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specifications
Product Family Options
Product Information
- PhaseCapacityBed weightAvailabilityPrice
- VARI12102158Straight barrel cartridge NH2Specifications:
More
Specifications
Cat. No.VARI12102158Carbon loadEndcappedSurface area - VARI12272005Straight barrel cartridge PAHSpecifications:
More
Specifications
Cat. No.VARI12272005Carbon load18,0%EndcappedNoSurface area500 m²/g - VARI12272004Straight barrel cartridge PesticideSpecifications:
More
Specifications
Cat. No.VARI12272004Carbon load18,0%EndcappedNoSurface area500 m²/g - VARI12272001Straight barrel cartridge oil and greaseSpecifications:
More
Specifications
Cat. No.VARI12272001Carbon load18,0%EndcappedNoSurface area500 m²/g - VARI12272011Straight barrel cartridge W/20ml Aquasep CeliteStraight barrel cartridge W/20ml Aquasep CeliteEnvirElut20 ml1 gLog in for availability1 * 20 ST348,84 €Specifications:
More
Specifications
Cat. No.VARI12272011Carbon load18,0%EndcappedNoSurface area500 m²/g - VARI12272020Straight barrel cartridge US EPA 1664Specifications:
More
Specifications
Cat. No.VARI12272020Carbon load18,0%EndcappedNoSurface area500 m²/g