Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ DEA
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ DEA
Supplier:  VARIAN
Certificates
11 Options Available Below
Product Details & Documents
Bond Elut™ DEA bears some resemblance to Bond Elut™ NH₂ in its properties but with a slightly lower capacity as an anion exchange sorbent. DEA has a moderately non-polar character due to the alkyl side chains on the amino functionality. These groups still afford a medium level of polarity, higher than C₈ but less polar than C₂ or CN-E.
  • More polar than C₈ but less polar than C₂ or CN
  • Lower ionic capacity than NH₂
  • Alkyl side chains confer moderately non-polar chracteristics
Bond Elut™ DEA is an excellent phase for the extraction of bisphosphonates from human serum and urine. Bisphosphonates are extremely hydrophilic and structurally similar to many endogenous phosphorylated compounds, making their selective extraction from serum or urine very challenging. The unique chemistry of Bond Elut™ DEA offers excellent selectivity of this class of compounds.
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113016
  • VARI12113042
  • VARI14113042
  • VARI12102043
  • VARI12102016
  • VARI14102078
  • VARI14102016
  • VARI14102043
  • VARI102019
  • VARI12102078
  • VARI12166046B
  • Carbon load:
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113016
    LRC cartridge
    LRC cartridge
    DEA
    60 Å
    40 µm
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113016
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12113042
    LRC cartridge
    LRC cartridge
    DEA
    60 Å
    40 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113042
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14113042
    LRC cartridge
    LRC cartridge
    DEA
    60 Å
    120 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 280,30 €
    280,30 €
    1 * 50 ST
       280,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14113042
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102043
    Straight barrel cartridge
    Straight barrel cartridge
    DEA
    60 Å
    120 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102043
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102016
    Straight barrel cartridge
    Straight barrel cartridge
    DEA
    60 Å
    40 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 346,80 €
    346,80 €
    1 * 100 ST
       346,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102016
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102078
    Straight barrel cartridge
    Straight barrel cartridge
    DEA
    60 Å
    120 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 337,59 €
    337,59 €
    1 * 100 ST
       337,59 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102078
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102016
    Straight barrel cartridge
    Straight barrel cartridge
    DEA
    60 Å
    40 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 346,80 €
    346,80 €
    1 * 100 ST
       346,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102016
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102043
    Straight barrel cartridge
    Straight barrel cartridge
    DEA
    60 Å
    40 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 301,78 €
    301,78 €
    1 * 50 ST
       301,78 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102043
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI102019
    Straight barrel cartridge
    Straight barrel cartridge
    DEA
    60 Å
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI102019
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102078
    Straight barrel cartridge
    Straight barrel cartridge
    DEA
    60 Å
    40 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 337,59 €
    337,59 €
    1 * 100 ST
       337,59 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102078
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g
Bond Elut™ Jr
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12166046B
    Bond Elut™ Jr
    Bond Elut™ Jr
    DEA
    60 Å
    40 µm
    1000 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 463,42 €
    463,42 €
    1 * 100 ST
       463,42 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12166046B
    Carbon load
    8,5%
    Endcapped
    No
    Surface area
    500 m²/g