Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ Certify
Solid phase extraction cartridges, Bond Elut™ Certify
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ Certify
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ Certify
33 Options Available Below
Product Details & Documents
The Bond Elut™ Certify extraction cartridge utilises a packed bed consisting of a special, non-polar C8 sorbent and a strong cation exchanger (SCX). Certify is most commonly used to extract basic (cationic) drugs from urine and blood, but it is also very effective for extraction of a wide range of compounds from a diverse range of aqueous matrices.
  • Special mixed mode sorbent bed
  • Broad application range for aqueous extraction
  • Bimodal, non-polar and strong cation exchange
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12105030
  • 551-0326
  • VARI12102051T
  • VARI52102051
  • VARI12256146
  • VARI12256146TJ
  • VARI12102145
  • VARI12256145
  • VARI52113051
  • VARI12102081
  • VARI12102081T
  • VARI52102081
  • VARI12102082
  • VARI12102093
  • VARI12102085
  • VARI14102083
  • VARI14102051
  • VARI14102081T
  • VARI14102093
  • VARI14102085
  • 551-0350
  • VARI52113050
  • VARI12113054
  • 551-0351
  • VARI14113050
  • VARI14113055
  • VARI14113054
  • VARI14113052
  • 551-0433
  • 551-0432
  • VARI75509025
  • VARI75509050
  • VARI7550901C
  • Carbon load:
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • 9,0%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12105030
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    3 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 197,44 €
    197,44 €
    1 * 50 ST
       197,44 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12105030
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • 551-0326
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    3 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    551-0326
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102051T
    Straight barrel cartridge, tabless
    Straight barrel cartridge, tabless
    Certify
    60 Å
    40 µm
    3 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 212,78 €
    212,78 €
    1 * 50 ST
       212,78 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102051T
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI52102051
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    3 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 657,26 €
    1 657,26 €
    1 * 500 ST
       1 657,26 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI52102051
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12256146
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    6 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 154,47 €
    154,47 €
    1 * 30 ST
       154,47 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256146
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12256146TJ
    Straight barrel cartridge, tabless
    Straight barrel cartridge, tabless
    Certify
    60 Å
    40 µm
    6 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 2 496,12 €
    2 496,12 €
    1 * 500 ST
       2 496,12 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256146TJ
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102145
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 212,78 €
    212,78 €
    1 * 50 ST
       212,78 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102145
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12256145
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    6 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 131,97 €
    131,97 €
    1 * 30 ST
       131,97 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256145
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI52113051
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    10 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 974,39 €
    1 974,39 €
    1 * 500 ST
       1 974,39 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI52113051
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102081
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    3 ml
    300 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102081
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102081T
    Straight barrel cartridge, tabless
    Straight barrel cartridge, tabless
    Certify
    60 Å
    40 µm
    3 ml
    300 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 217,90 €
    217,90 €
    1 * 50 ST
       217,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102081T
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI52102081
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    3 ml
    300 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 657,26 €
    1 657,26 €
    1 * 500 ST
       1 657,26 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI52102081
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102082
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    6 ml
    300 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102082
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102093
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 192,32 €
    192,32 €
    1 * 30 ST
       192,32 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102093
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12102085
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    40 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 237,34 €
    237,34 €
    1 * 30 ST
       237,34 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102085
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102083
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    120 µm
    1 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 402,04 €
    402,04 €
    1 * 100 ST
       402,04 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102083
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102051
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    120 µm
    3 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 234,27 €
    234,27 €
    1 * 50 ST
       234,27 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102051
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102081T
    Straight barrel cartridge, tabless
    Straight barrel cartridge, tabless
    Certify
    60 Å
    120 µm
    3 ml
    300 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 203,58 €
    203,58 €
    1 * 50 ST
       203,58 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102081T
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102093
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    120 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 212,78 €
    212,78 €
    1 * 30 ST
       212,78 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102093
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14102085
    Straight barrel cartridge
    Straight barrel cartridge
    Certify
    60 Å
    120 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 233,24 €
    233,24 €
    1 * 30 ST
       233,24 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102085
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
LRC cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 551-0350
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    40 µm
    10 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    551-0350
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI52113050
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    40 µm
    10 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 657,26 €
    1 657,26 €
    1 * 500 ST
       1 657,26 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI52113050
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI12113054
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    40 µm
    10 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 223,01 €
    223,01 €
    1 * 50 ST
       223,01 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113054
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • 551-0351
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    40 µm
    10 ml
    300 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    551-0351
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14113050
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    120 µm
    10 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 210,74 €
    210,74 €
    1 * 50 ST
       210,74 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14113050
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14113055
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    120 µm
    10 ml
    130 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 647,03 €
    1 647,03 €
    1 * 500 ST
       1 647,03 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14113055
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14113054
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    120 µm
    10 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 219,95 €
    219,95 €
    1 * 50 ST
       219,95 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14113054
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI14113052
    LRC cartridge
    LRC cartridge
    Certify
    60 Å
    120 µm
    10 ml
    300 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 227,11 €
    227,11 €
    1 * 50 ST
       227,11 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14113052
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
VersaPlate™ formats
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 551-0433
    VersaPlate™ preassembled 96-well plate
    VersaPlate™ preassembled 96-well plate
    Certify
    60 Å
    40 µm
    40 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    551-0433
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • 551-0432
    VersaPlate™ preassembled 96-well plate
    VersaPlate™ preassembled 96-well plate
    Certify
    60 Å
    40 µm
    40 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 586,18 €
    586,18 €
    1 * 1 ST
       586,18 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0432
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI75509025
    VersaPlate™ tube
    VersaPlate™ tube
    Certify
    60 Å
    40 µm
    40 ml
    25 mg
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 414,31 €
    414,31 €
    1 * 96 ST
       414,31 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI75509025
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI75509050
    VersaPlate™ tube
    VersaPlate™ tube
    Certify
    60 Å
    40 µm
    40 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 432,73 €
    432,73 €
    1 * 96 ST
       432,73 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI75509050
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g
  • VARI7550901C
    VersaPlate™ tube
    VersaPlate™ tube
    Certify
    60 Å
    40 µm
    40 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 432,73 €
    432,73 €
    1 * 96 ST
       432,73 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI7550901C
    Carbon load
    9,0%
    Endcapped
    No
    Surface area
    500 m²/g