Graduated pipettes, with piston
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
About this item
Safety measuring pipettes with interchangeable ground-in piston, standard type.
- Tolerance 1,5 times higher than the official tolerance of class AS
- Fused amber graduation 'ASSISTENT-PERMANENT'
- Zero at top, calibrated
- Fire-polished tip
: Each pipette is packed in a plastic tube.
6 Options Available Below
Product Details & Documents
Safety measuring pipettes with interchangeable ground-in piston, standard type.
- Tolerance 1,5 times higher than the official tolerance of class AS
- Fused amber graduation 'ASSISTENT-PERMANENT'
- Zero at top, calibrated
- Fire-polished tip
: Each pipette is packed in a plastic tube.
Product Family Options
Product Information
- KapazitätTeilungToleranzFarbcodeAvailabilityPrice