ContactUsLinkComponent
HPLC columns, Acclaim™ mixed-mode HILIC-1
HPLC columns, Acclaim™ mixed-mode HILIC-1
Certificates
undefined
HPLC columns, Acclaim™ mixed-mode HILIC-1
Specifications
  • Catalog No:
  • 554-4456
  • 554-4455
  • 554-4461
  • 554-4457
  • 555-7128
  • 555-7138
  • 554-4460
  • 554-4462
  • 555-7170
  • Max. pressure:
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • 8500 psi (4800 psi for 3,0×50 mm column)
  • pH range:
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • Temperature limit:
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C