Orders LinkContactUsLinkComponent
Filter forceps
Filter forceps
Straight, blunt
Catalog # MILFXX6200006P
Supplier:   Merck
Filter forceps
Catalog # MILFXX6200006P
Supplier:  Merck
Filter forceps
Filter forceps
Straight, blunt
Price
78,10 €
Log in for availability
1 * 3 ST
Product Details & Documents
Sterilisable filter forceps made from stainless steel, with PP covered handles (in three different colours).
Specifications
  • Type:
    Straight, blunt