Order Entry
Austria
ContactUsLinkComponent
pH/mV/°C meters, bench, SevenExcellence™ S400
pH/mV/°C meters, bench, SevenExcellence™ S400
Supplier:  Mettler - Toledo
Certificates
undefined
pH/mV/°C meters, bench, SevenExcellence™ S400
Supplier:  Mettler - Toledo

Specifications

  • Catalog No:
  • 662-2000
  • 662-1466
  • 662-1464
  • 662-1467
  • Model:
  • S400
  • S400
  • S400
  • S400
  • pH range:
  • –2...+20
    ISFET pH: 0...14
  • –2...+20
    ISFET pH: 0...14
  • –2...+20
    ISFET pH: 0...14
  • –2...+20
    ISFET pH: 0...14
  • pH resolution:
  • 0,001/0,01/0,1
  • 0,001/0,01/0,1
  • 0,001/0,01/0,1
  • 0,001/0,01/0,1
  • pH accuracy:
  • ±0,002
    ISFET pH: ±0,05
  • ±0,002
    ISFET pH: ±0,05
  • ±0,002
    ISFET pH: ±0,05
  • ±0,002
    ISFET pH: ±0,05
  • pH calibration:
  • Max. 5 points
  • Max. 5 points
  • Max. 5 points
  • Max. 5 points
  • Automatic buffer recognition:
  • Automatic or manual, 8 pre-defined and 20 user-defined buffer groups
  • Automatic or manual, 8 pre-defined and 20 user-defined buffer groups
  • Automatic or manual, 8 pre-defined and 20 user-defined buffer groups
  • Automatic or manual, 8 pre-defined and 20 user-defined buffer groups
  • mV range:
  • ±2000
  • ±2000
  • ±2000
  • ±2000
  • mV resolution:
  • 0,1/1
  • 0,1/1
  • 0,1/1
  • 0,1/1
  • mV accuracy:
  • ±0,1
  • ±0,1
  • ±0,1
  • ±0,1
  • Temperature range:
  • −30...+130 °C
  • −30...+130 °C
  • −30...+130 °C
  • −30...+130 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Display:
  • TFT colour display
  • TFT colour display
  • TFT colour display
  • TFT colour display
  • Compliance:
  • GLP
  • GLP
  • GLP
  • GLP
  • Data logging capacity:
  • 20000 data points, 250 analyses
  • 20000 data points, 250 analyses
  • 20000 data points, 250 analyses
  • 20000 data points, 250 analyses
  • Connections:
  • BNC, NTC30 kΩ temperature sensor input, Mini-DIN ISFET sensor input, Mini-LTW digital sensor input, Mini-DIN stirrer connection
  • BNC, NTC30 kΩ temperature sensor input, Mini-DIN ISFET sensor input, Mini-LTW digital sensor input, Mini-DIN stirrer connection
  • BNC, NTC30 kΩ temperature sensor input, Mini-DIN ISFET sensor input, Mini-LTW digital sensor input, Mini-DIN stirrer connection
  • BNC, NTC30 kΩ temperature sensor input, Mini-DIN ISFET sensor input, Mini-LTW digital sensor input, Mini-DIN stirrer connection
  • Outputs:
  • USB, RS232, LAN
  • USB, RS232, LAN
  • USB, RS232, LAN
  • USB, RS232, LAN
  • W×D×H:
  • 235×188×75 mm
  • 235×188×75 mm
  • 235×188×75 mm
  • 235×188×75 mm
  • Weight:
  • 1,12 kg
  • 1,12 kg
  • 1,12 kg
  • 1,12 kg

Specifications

Related Information

Frequently Bought Together