PCR tube racks, PP, Axygen®
: Corning
PP with lid.
- Fluorescent colour base with translucent lid
- Durable, chemically inert, and can withstand extended duration at sub-zero temperatures
Designed to hold 0,2, and 0,5 ml PCR tubes.
: Assorted pack includes one each of blue, green, violet, red and yellow.
- Catalog No:
- 732-1389
- 732-1388
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Frequently Bought Together