Order Entry
Austria
ContactUsLinkComponent
 
Reagent reservoirs, Labcor™
Catalog # BITX730-004
Supplier:  BIOTIX
undefined
Reagent reservoirs, Labcor™
Catalog # BITX730-004
Supplier:  BIOTIX

Specifications

  • Packed:
    80/case (individually wrapped)
  • Sterile:
    +
  • Type:
    PS, V-shaped
  • Capacity:
    55 ml
  • Seq:
    02
  • W×D×H:
    143×7×25 mm
  • Pk:
    80

Specifications

About this item

PS or PVC disposable reservoirs are designed for 8- and 12-channel pipettors.

  • Angled sides and V-shaped bottom permit easy access to solution
  • Available sterile (PS) or non sterile (PVC)
  • Sterile reservoirs are electron beam sterilised and RNase-, DNase-, and pyrogen-free
  • PVC material allows the contents to be clearly viewed