Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ C2
Solid phase extraction cartridges, Bond Elut™ C2
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ C2
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ C2
Solid phase extraction cartridges, Bond Elut™ C2
10 Options Available Below
Product Details & Documents
Bond Elut™ C2 is a fairly non-polar sorbent because of the short chain length of the functional group. C2 is often used during the process of method development if analytes are retained too strongly on a C8 or C18 phase. The polarity of C2 is slightly lower than a cyano phase for polar interactions.
  • C2 is a low carbon load sorbent
  • Can be used alongside CN and C8 phases
  • Popular for drug extraction from plasma and for flat baselines
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113003
  • VARI12113029
  • VARI12102060
  • VARI12105029
  • 551-0302
  • VARI12102117
  • 551-0314
  • 551-0317
  • VARI12102115
  • VARI12256003
  • Carbon load:
  • 5,6%
  • 5,6%
  • 5,6%
  • 5,6%
  • 5,6%
  • 5,6%
  • 5,6%
  • 5,6%
  • 5,6%
  • 5,6%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113003
    LRC cartridge
    LRC cartridge
    C2
    60 Å
    40 µm
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 163,68 €
    163,68 €
    1 * 50 ST
       163,68 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113003
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12113029
    LRC cartridge
    LRC cartridge
    C2
    60 Å
    40 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 244,50 €
    244,50 €
    1 * 50 ST
       244,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113029
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102060
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 285,42 €
    285,42 €
    1 * 100 ST
       285,42 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102060
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12105029
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    3 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 179,02 €
    179,02 €
    1 * 50 ST
       179,02 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12105029
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0302
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 273,14 €
    273,14 €
    1 * 100 ST
       273,14 €
    Specifications:
    More
    Specifications
    Cat. No.
    551-0302
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12102117
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    3 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 187,21 €
    187,21 €
    1 * 50 ST
       187,21 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102117
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0314
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    551-0314
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • 551-0317
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    551-0317
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12102115
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 132,99 €
    132,99 €
    1 * 30 ST
       132,99 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102115
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g
  • VARI12256003
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    60 Å
    40 µm
    6 ml
    1 g
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 200,51 €
    200,51 €
    1 * 30 ST
       200,51 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12256003
    Carbon load
    5,6%
    Endcapped
    Yes
    Surface area
    500 m²/g