ContactUsLinkComponent
Multi-position magnetic stirrers, MultiMagStir Genie®
Multi-position magnetic stirrers, MultiMagStir Genie®
Certificates
Multi-position magnetic stirrers, MultiMagStir Genie®
Specifications
  • Catalog No:
  • SCIISI-3002
  • Model:
  • MultiMagStir Genie®
  • No. of stirring positions:
  • 4
  • Max. stirring capacity per stirrer H₂O:
  • 1000 ml ×4 (or max 3×1 L spinner flasks)
  • Speed range:
  • 10 - 1200 min⁻¹
  • Plate material:
  • ABS/PC
  • W×D×H:
  • 325×375×48 mm
  • Weight:
  • 3,2 kg