Menadione (2-Methyl-1,4-naphthoquinone, Vitamin K3)
Catalog # EHERC14863000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:172.19 g/mol
EINECS:200-372-6
CAS Number:58-27-5
Formula:C₁₁H₈O₂
MDL Number:MFCD00001681