Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiophen-2-acetonitrile
Thiophen-2-acetonitrile, Sigma-Aldrich®
Thiophen-2-acetonitrile, Sigma-Aldrich®
Thiophen-2-acetonitrile, Sigma-Aldrich®
SIAL141682-10G
:
Merck
:
Thiophen-2-acetonitrile, Sigma-Aldrich®
SIAL141682-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
6.1,III
:
M-Clarity™ quality level = MQ200
:
123.18 g/mol
:
244-104-6
:
3276
:
20893-30-5
:
C₆H₅NS
:
MFCD00005453