Order Entry
Austria
ContactUsLinkComponent
L(+)-Glutamine 0.2 mol/l in aqueous solution
L(+)-Glutamine 0.2 mol/l in aqueous solution
Artikelnr. X0550-100
Supplier:  Biowest
CAS Number:  
undefined
L(+)-Glutamine 0.2 mol/l in aqueous solution
Artikelnr. X0550-100
Supplier:  Biowest
CAS Number:  

Specifications

  • Pk:
    100 ml
  • Seq:
    2440

Specifications

About this item

 :
 :146.15 g/mol
Merck Index:13,04484
Storage Temperature:Ambient
EINECS:200-292-1
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044