Order Entry
Austria
ContactUsLinkComponent
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥95% (by perchloric acid titration) for fluorescence, Sigma-Aldrich®
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥95% (by perchloric acid titration) for fluorescence, Sigma-Aldrich®
Certificates
undefined
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥95% (by perchloric acid titration) for fluorescence, Sigma-Aldrich®

Specification Test Results

  • Appearance (colour):
    Faint green to green
  • Appearance (form):
    Powder
  • Titration (nt) HClO₄ 0.1 m:
    Anhydrous basis
  • Purity (TLC area %):
    ≥ 95 %
  • Solubility (colour):
    Green to very dark green and brown to very dark brown
  • Solubility (turbidity):
    Clear to very hazy
  • Solubility (method):
    50 mg/ml in water
  • Water:
    Report result
  • Recommended retest period:
    36 months

Specification Test Results

Frequently Bought Together