Order Entry
Austria
ContactUsLinkComponent
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥95%, Sigma-Aldrich®
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥95%, Sigma-Aldrich®
Certificates
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥95%, Sigma-Aldrich®

Specification Test Results

  • Appearance (colour):
    Faint green to green
  • Appearance (form):
    Powder
  • Titration (NT) HClO₄ 0.1 m:
    ≥95.0%
  • Purity (TLC area %):
    ≥95.0 %
  • Solubility (colour):
    Green to very dark green and brown to very dark brown
  • Solubility (turbidity):
    Clear to very hazy
  • Solubility (method):
    50 mg/ml water, hot
  • Water:
    5.0 - 8.0%
  • Carbon content:
    58.50% (theory)
  • Hydrogen content:
    4.30% (theory)
  • Nitrogen content:
    4.26% (theory)
  • Infrared spectrum:
    Conforms to structure
  • Emission maximum:
    465 - 475 nm
  • Remarks on luminescence:
    Exc.:388 nm 0.1 M tris; 0.2 mkcl; ph 9.0; BSA
  • Recommended retest period:
    36 months

Specification Test Results

Frequently Bought Together