Order Entry
Austria
ContactUsLinkComponent
Progesterone, Sigma-Aldrich®
Progesterone, Sigma-Aldrich®
Catalog # SIALP6149-1MG
Supplier:  Merck
CAS Number:  
Progesterone, Sigma-Aldrich®
Catalog # SIALP6149-1MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
EINECS:200-350-6
CAS Number:57-83-0
Formula:C₂₁H₃₀O₂
MDL Number:MFCD00003658