Progesterone
Catalog # FITZ30-AP04
Supplier: FITZGERALD INDUSTRIES INTL
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
EINECS:200-350-6
CAS Number:57-83-0
Formula:C₂₁H₃₀O₂
MDL Number:MFCD00003658